Tisk…

(2-Methyl-1,3-thiazol-4-yl)methanol 97%

Dodavatel: Apollo Scientific

Synonyms: (2-Methylthiazol-4-yl)methanol

OR962726-25G OR962726-5G
APOSOR962726-25GEA 17130 CZK
APOSOR962726-25G APOSOR962726-5G
(2-Methyl-1,3-thiazol-4-yl)methanol 97%
(2-Methyl-1,3-thiazol-4-yl)methanol
Vzorec: C₅H₇NOS
MW: 129,1802 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08703637
CAS číslo: 76632-23-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR