Tisk…

3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]benzoic acid 96%

Dodavatel: Apollo Scientific

Synonyms: 3-(4-Piperazin-1-yl)benzoic acid, N-BOC protected, 3-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid

OR962653-1G OR962653-5G
APOSOR962653-1GEA 4930 CZK
APOSOR962653-1G APOSOR962653-5G
3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]benzoic acid 96%
3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]benzoic acid
Vzorec: C₁₆H₂₂N₂O₄
MW: 306,36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06657287
CAS číslo: 193818-13-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR