Tisk…

3-(4-Chlorophenoxy)propylamine 97%

Dodavatel: Apollo Scientific

OR962649-1G OR962649-5G
APOSOR962649-1GEA 6220 CZK
APOSOR962649-1G APOSOR962649-5G
3-(4-Chlorophenoxy)propylamine 97%
3-(4-Chlorophenoxy)propylamine
Vzorec: C₉H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD06213576
CAS číslo: 50911-60-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR