1-(2-nitrophenyl)pyrrolidine 98%
Dodavatel: Apollo Scientific
OR962562-100G
OR962562-25G
OR962562-5G
APOSOR962562-100GEA
17990
CZK
APOSOR962562-100G
APOSOR962562-25G
APOSOR962562-5G
1-(2-nitrophenyl)pyrrolidine 98%
1-(2-nitrophenyl)pyrrolidine
Vzorec:
C₁₀H₁₂N₂O₂ MW: 192,218 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01604393 CAS číslo: 40832-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...