Tisk…

4-(Hydroxymethyl)benzonitrile 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Cyanobenzyl alcohol

OR962541-25G OR962541-100G
APOSOR962541-25GEA 2900 CZK
APOSOR962541-25G APOSOR962541-100G
4-(Hydroxymethyl)benzonitrile 98%
4-(Hydroxymethyl)benzonitrile
Vzorec: C₈H₇NO
MW: 133.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00870633
CAS číslo: 874-89-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR