Tisk…

4-Acetoxybenzonitrile 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Cyanophenyl acetate

OR962477-25G OR962477-5G
APOSOR962477-25GEA 9720 CZK
APOSOR962477-25G APOSOR962477-5G
4-Acetoxybenzonitrile 98%
4-Acetoxybenzonitrile
Vzorec: C₉H₇NO₂
MW: 161,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129158
CAS číslo: 13031-41-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR