N-Isopropyl-4-nitroaniline 95%
Dodavatel: Apollo Scientific
Synonyms:
N-isopropyl-4-nitroaniline
OR962476-1G
OR962476-5G
APOSOR962476-1GEA
1040
CZK
APOSOR962476-1G
APOSOR962476-5G
N-Isopropyl-4-nitroaniline 95%
N-Isopropyl-4-nitroaniline
Vzorec:
C₉H₁₂N₂O₂ MW: 180,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00128997 CAS číslo: 25186-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...