Tisk…

Ethyl-2,4-dimethylnicotinate 97%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2,4-dimethylpyridine-3-carboxylate, Ethyl 2,4-dimethylnicotinate

OR962464-25G OR962464-5G
APOSOR962464-25GEA 9230 CZK
APOSOR962464-25G APOSOR962464-5G
Ethyl-2,4-dimethylnicotinate 97%
Ethyl-2,4-dimethylnicotinate
Vzorec: C₁₀H₁₃NO₂
MW: 179,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051962
CAS číslo: 37669-78-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR