(±)-4-Chlorostyrene oxide 97%
Dodavatel: Apollo Scientific
Synonyms:
2-(4-Chlorophenyl)oxirane
OR962433-1G
OR962433-5G
APOSOR962433-1GEA
3310
CZK
APOSOR962433-1G
APOSOR962433-5G
(±)-4-Chlorostyrene oxide 97%
(±)-4-Chlorostyrene oxide
Vzorec:
C₈H₇ClO MW: 154.6 Hustota: 1.283 g/cm³ (25 °C) Bod vzplanutí: 20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00956179 CAS číslo: 2788-86-5 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...