Tisk…

N-Boc-3-cyano-L-phenylalanine 97%

Dodavatel: Apollo Scientific

Synonyms: Boc-Phe(3-CN)-OH

OR962365-5G
APOSOR962365-5GEA 2110 CZK
APOSOR962365-5G
N-Boc-3-cyano-L-phenylalanine 97%
N-Boc-3-cyano-L-phenylalanine
Vzorec: C₁₅H₁₈N₂O₄
MW: 290,319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00797560
CAS číslo: 131980-30-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR