Fmoc-Nα-methyl-L-phenylglycine 97%
Dodavatel: Apollo Scientific
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-2-phenylacetic acid, N-Fmoc-(S)-a-(methylamino)-benzeneacetic acid, Fmoc-N-Me-Phg-OH
OR962347-1G
OR962347-5G
APOSOR962347-1GEA
3540
CZK
APOSOR962347-1G
APOSOR962347-5G
Fmoc-Nα-methyl-L-phenylglycine 97%
Fmoc-Nalpha-methyl-L-phenylglycine
Vzorec:
C₂₄H₂₁NO₄ MW: 387.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01861296 CAS číslo: 574739-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...