Tisk…

Fmoc-N-methyl-L-norvaline 97%

Dodavatel: Apollo Scientific

Synonyms: Fmoc-N-Me-Nva-OH, N-Fmoc-N-methyl-L-norvaline, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)pentanoic acid

OR962325-500MG OR962325-5G OR962325-25G
APOSOR962325-500MGEA 0 CZK
APOSOR962325-500MG APOSOR962325-5G APOSOR962325-25G
Fmoc-N-methyl-L-norvaline 97%
Fmoc-N-methyl-L-norvaline
Vzorec: C₂₁H₂₃NO₄
MW: 353,418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861326
CAS číslo: 252049-05-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR