Tisk…

H-2-Abu-OtBu·HCl 98%

Dodavatel: Apollo Scientific

Synonyms: H-Abu-OtBu.HCl, tert-Butyl (S)-2-aminobutyrate hydrochloride, H-Abu-OtBu·HCl

OR962286-100G OR962286-25G OR962286-5G
APOSOR962286-100GEA 7210 CZK
APOSOR962286-100G APOSOR962286-25G APOSOR962286-5G
H-2-Abu-OtBu·HCl 98%
H-2-Abu-OtBu·HCl
Vzorec: C₈H₁₈ClNO₂
MW: 195,687 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237697
CAS číslo: 53956-05-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR