DDe-OH 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(1-Hydroxyethylidene)-5,5-dimethylcyclohexane-1,3-dione
OR962225-100G
OR962225-25G
OR962225-5G
APOSOR962225-100GEA
11150
CZK
APOSOR962225-100G
APOSOR962225-25G
APOSOR962225-5G
DDe-OH 98%
DDe-OH
Vzorec:
C₁₀H₁₄O₃ MW: 182.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03001669 CAS číslo: 94142-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...