Phenyl-1-Propenyl Ketone 90%
Dodavatel: Apollo Scientific
Synonyms:
1-Phenyl-2-buten-1-one
OR962116-1G
OR962116-25G
OR962116-5G
APOSOR962116-1GEA
3540
CZK
APOSOR962116-1G
APOSOR962116-25G
APOSOR962116-5G
Phenyl-1-Propenyl Ketone 90%
Phenyl-1-Propenyl Ketone
Vzorec:
C₁₀H₁₀O MW: 146,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059386 CAS číslo: 495-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...