Fmoc-3-cyano-L-phenylalanine 97%
Dodavatel: Apollo Scientific
Synonyms:
Fmoc-Phe(3-CN)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-cyanophenyl)propanoic acid
OR961964-5G
APOSOR961964-5GEA
3580
CZK
APOSOR961964-5G
Fmoc-3-cyano-L-phenylalanine 97%
Fmoc-3-cyano-L-phenylalanine
Vzorec:
C₂₅H₂₀N₂O₄ MW: 412,45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00797585 CAS číslo: 205526-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...