Tisk…

1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile 95%

Dodavatel: Apollo Scientific

Synonyms: 6-Cyano-7-azaindole

OR961921-1G OR961921-250MG
APOSOR961921-1GEA 7840 CZK
APOSOR961921-1G APOSOR961921-250MG
1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile 95%
1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile
Vzorec: C₈H₅N₃
MW: 143,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659673
CAS číslo: 189882-33-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR