Tisk…

1-Benzyl-1H-pyrazole-4-carbonitrile 95%

Dodavatel: Apollo Scientific

OR961625-1G OR961625-250MG
APOSOR961625-1GEA 12400 CZK
APOSOR961625-1G APOSOR961625-250MG
1-Benzyl-1H-pyrazole-4-carbonitrile 95%
1-Benzyl-1H-pyrazole-4-carbonitrile
Vzorec: C₁₁H₉N₃
Storage Temperature: Ambient
CAS číslo: 121358-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR