Methyl-1-aminocyclobutanecarboxylate hydrochloride
Dodavatel: Apollo Scientific
Synonyms:
Methyl 1-aminocyclobutanecarboxylate hydrochloride
OR961598-25G
OR961598-5G
APOSOR961598-25GEA
12240
CZK
APOSOR961598-25G
APOSOR961598-5G
Methyl-1-aminocyclobutanecarboxylate hydrochloride
Methyl-1-aminocyclobutanecarboxylate hydrochloride
Vzorec:
C₆H₁₂ClNO₂ MW: 165,617 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04115288 CAS číslo: 92398-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...