1-(Thiazol-2-yl)ethanamine hydrochloride 95%
Dodavatel: Apollo Scientific
OR961487-1G
OR961487-250MG
APOSOR961487-1GEA
2570
CZK
APOSOR961487-1G
APOSOR961487-250MG
1-(Thiazol-2-yl)ethanamine hydrochloride 95%
1-(Thiazol-2-yl)ethanamine hydrochloride
Vzorec:
C₅H₉ClN₂S Storage Temperature: Ambient |
MDL Number:
MFCD11846195 CAS číslo: 947662-64-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...