Tisk…

N-Me-Ile-OMe·HCl 97%

Dodavatel: Apollo Scientific

Synonyms: N-Me-Ile-OMe·HCl, (2S,3S)-Methyl 3-methyl-2-(methylamino)pentanoate hydrochloride

OR961408-1G OR961408-250MG
APOSOR961408-1GEA 8240 CZK
APOSOR961408-1G APOSOR961408-250MG
N-Me-Ile-OMe·HCl 97%
N-Me-Ile-OMe·HCl
Vzorec: C₈H₁₈ClNO₂
MW: 195,687 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093476
CAS číslo: 3339-43-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR