Tisk…

Boc-N-ME-NVA-OH 97%

Dodavatel: Apollo Scientific

Synonyms: Boc-N-methyl-L-norvaline, (S)-2-((tert-Butoxycarbonyl)(methyl)amino)pentanoic acid

OR961348-1G OR961348-5G
APOSOR961348-1GEA 4860 CZK
APOSOR961348-1G APOSOR961348-5G
Boc-N-ME-NVA-OH 97%
Boc-N-ME-NVA-OH
Vzorec: C₁₁H₂₁NO₄
MW: 231,292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094111
CAS číslo: 136092-78-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR