Tisk…

Boc-L-Asp(OAll)-OH

Dodavatel: Apollo Scientific

Synonyms: Boc-Asp(OAll)-OH, (S)-4-(Allyloxy)-2-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid

OR961039-25G OR961039-5G OR961039-100G
APOSOR961039-25GEA 8600 CZK
APOSOR961039-25G APOSOR961039-5G APOSOR961039-100G
Boc-L-Asp(OAll)-OH
Boc-L-Asp(OAll)-OH
Vzorec: C₁₂H₁₉NO₆
Bod tání: 44…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00273398
CAS číslo: 132286-77-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR