N-Me-Val-OMe·HCl 95%
Dodavatel: Apollo Scientific
OR961005-1G
OR961005-250MG
APOSOR961005-1GEA
662
CZK
APOSOR961005-1G
APOSOR961005-250MG
N-Me-Val-OMe·HCl 95%
N-Me-Val-OMe·HCl
Vzorec:
C₇H₁₆ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00238325 CAS číslo: 3339-44-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...