6-Ethoxyimidazo[1,2-b]pyridazine 95%
Dodavatel: Apollo Scientific
OR960828-1G
OR960828-250MG
APOSOR960828-1GEA
10350
CZK
APOSOR960828-1G
APOSOR960828-250MG
6-Ethoxyimidazo[1,2-b]pyridazine 95%
6-Ethoxyimidazo[1,2-b]pyridazine
Vzorec:
C₈H₉N₃O MW: 163.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18782899 CAS číslo: 57470-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...