Tisk…

3-Ethoxy-2,2-dimethylcyclobutanone 95%

Dodavatel: Apollo Scientific

OR960827-1G OR960827-250MG
APOSOR960827-1GEA 22950 CZK
APOSOR960827-1G APOSOR960827-250MG
3-Ethoxy-2,2-dimethylcyclobutanone 95%
3-Ethoxy-2,2-dimethylcyclobutanone
Vzorec: C₈H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD18452159
CAS číslo: 2292-84-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR