Tisk…

N-Fmoc-N-methyl-D-phenylalanine 97%

Dodavatel: Apollo Scientific

OR960818-1G OR960818-500MG OR960818-5G
APOSOR960818-1GEA 3580 CZK
APOSOR960818-1G APOSOR960818-500MG APOSOR960818-5G
N-Fmoc-N-methyl-D-phenylalanine 97%
N-Fmoc-N-methyl-D-phenylalanine
Vzorec: C₂₅H₂₃NO₄
MW: 401,462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153392
CAS číslo: 138775-05-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR