Tisk…

α,α-Dimethyl-2-thiazolemethanol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Thiazol-2-yl-propan-2-ol

OR960766-1G OR960766-250MG
APOSOR960766-1GEA 10350 CZK
APOSOR960766-1G APOSOR960766-250MG
α,α-Dimethyl-2-thiazolemethanol 95%
alpha,alpha-Dimethyl-2-thiazolemethanol
Vzorec: C₆H₉NOS
Storage Temperature: Ambient
MDL Number: MFCD03211903
CAS číslo: 16077-78-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR