S-(2-Chloroallyl)thioacetate 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethanethioic acid s-(2-chloro-2-propen-1-yl)ester
OR960695-1G
OR960695-5G
APOSOR960695-1GEA
6220
CZK
APOSOR960695-1G
APOSOR960695-5G
S-(2-Chloroallyl)thioacetate 95%
S-(2-Chloroallyl)thioacetate
Vzorec:
C₅H₇ClOS MW: 150,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18839251 CAS číslo: 24891-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...