6-Chloropyrido[3,2-b]pyrazine 98%
Dodavatel: Apollo Scientific
OR960682-1G
OR960682-250MG
APOSOR960682-1GEA
10550
CZK
APOSOR960682-1G
APOSOR960682-250MG
6-Chloropyrido[3,2-b]pyrazine 98%
6-Chloropyrido[3,2-b]pyrazine
Vzorec:
C₇H₄ClN₃ Storage Temperature: Ambient |
MDL Number:
MFCD11518952 CAS číslo: 68236-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...