Tisk…

Cyclopentylmethanamine hydrochloride 98%

Dodavatel: Apollo Scientific

OR960681-1G OR960681-250MG OR960681-5G
APOSOR960681-1GEA 7670 CZK
APOSOR960681-1G APOSOR960681-250MG APOSOR960681-5G
Cyclopentylmethanamine hydrochloride 98%
Cyclopentylmethanamine hydrochloride
Vzorec: C₆H₁₄ClN
Storage Temperature: Ambient
MDL Number: MFCD07403659
CAS číslo: 58714-85-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR