Tisk…

N-Fmoc-D-glutamic acid 5-tert-butyl ester 97%

Dodavatel: Apollo Scientific

Synonyms: 5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate, 5-tert-Butyl N-Fmoc-D-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 5-tert-Butyl Ester, Fmoc-D-Glu(OtBu)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid

OR960642-25G OR960642-100G
APOSOR960642-25GEA 1700 CZK
APOSOR960642-25G APOSOR960642-100G
N-Fmoc-D-glutamic acid 5-tert-butyl ester 97%
N-Fmoc-D-glutamic acid 5-tert-butyl ester
Vzorec: C₂₄H₂₇NO₆
Bod tání: 83…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077055
CAS číslo: 104091-08-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR