Tisk…

2-Chloro-4-(tributylstannyl)pyridine 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR960627-1G OR960627-5G
APOSOR960627-1GEA 4860 CZK
APOSOR960627-1G APOSOR960627-5G
2-Chloro-4-(tributylstannyl)pyridine 97%
2-Chloro-4-(tributylstannyl)pyridine
Vzorec: C₁₇H₃₀ClNSn
Storage Temperature: Ambient
MDL Number: MFCD10699154
CAS číslo: 1204580-73-3
UN: 2788
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR