3-Chloro-4-isopropoxycyclobutene-1,2-dione 95%
Dodavatel: Apollo Scientific
OR960613-1G
OR960613-250MG
OR960613-5G
APOSOR960613-1GEA
6090
CZK
APOSOR960613-1G
APOSOR960613-250MG
APOSOR960613-5G
3-Chloro-4-isopropoxycyclobutene-1,2-dione 95%
3-Chloro-4-isopropoxycyclobutene-1,2-dione
Vzorec:
C₇H₇ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09763707 CAS číslo: 130837-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...