Tisk…

Boc-N-methyl-D-leucine 97%

Dodavatel: Apollo Scientific

OR960580-25G OR960580-5G
APOSOR960580-25GEA 3940 CZK
APOSOR960580-25G APOSOR960580-5G
Boc-N-methyl-D-leucine 97%
Boc-N-methyl-D-leucine
Vzorec: C₁₂H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00076953
CAS číslo: 89536-84-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR