Tisk…

4-Bromo-1H-pyrazolo[3,4-b]pyridine 98%

Dodavatel: Apollo Scientific

OR960576-1G OR960576-250MG
APOSOR960576-1GEA 9720 CZK
APOSOR960576-1G APOSOR960576-250MG
4-Bromo-1H-pyrazolo[3,4-b]pyridine 98%
4-Bromo-1H-pyrazolo[3,4-b]pyridine
Vzorec: C₆H₄BrN₃
MW: 198,0201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699404
CAS číslo: 856859-49-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR