[1,2,4]Triazolo[1,5-a]pyridine 95%
Dodavatel: Apollo Scientific
OR960461-1G
OR960461-5G
APOSOR960461-1GEA
1490
CZK
APOSOR960461-1G
APOSOR960461-5G
[1,2,4]Triazolo[1,5-a]pyridine 95%
[1,2,4]Triazolo[1,5-a]pyridine
Vzorec:
C₆H₅N₃ Storage Temperature: Ambient |
MDL Number:
MFCD03700708 CAS číslo: 274-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...