Tisk…

2-(tert-Butyldimethylsilyl)thiazole 97%

Dodavatel: Apollo Scientific

OR960458-5G
APOSOR960458-5GEA 2030 CZK
APOSOR960458-5G
2-(tert-Butyldimethylsilyl)thiazole 97%
2-(tert-Butyldimethylsilyl)thiazole
Vzorec: C₉H₁₇NSSi
Storage Temperature: Ambient
MDL Number: MFCD10699169
CAS číslo: 137382-38-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR