3-(4-Aminophenyl)-N,N-dimethylpropanamide 98%
Dodavatel: Apollo Scientific
OR960347-1G
OR960347-5G
APOSOR960347-1GEA
7670
CZK
APOSOR960347-1G
APOSOR960347-5G
3-(4-Aminophenyl)-N,N-dimethylpropanamide 98%
3-(4-Aminophenyl)-N,N-dimethylpropanamide
Vzorec:
C₁₁H₁₆N₂O Storage Temperature: Ambient |
MDL Number:
MFCD10022146 CAS číslo: 1018284-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...