Tisk…

1,4-Dibromo-2-nitrobenzene 98%

Dodavatel: Apollo Scientific

Synonyms: 2,5-Dibromonitrobenzene

OR9603-100G OR9603-1KG OR9603-500G
APOSOR9603-100GEA 1990 CZK
APOSOR9603-100G APOSOR9603-1KG APOSOR9603-500G
1,4-Dibromo-2-nitrobenzene 98%
1,4-Dibromo-2-nitrobenzene
Vzorec: C₆H₃Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD00007046
CAS číslo: 3460-18-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR