Tisk…

6-nitro-3,4-dihydro-2H-1,4-benzoxazine 98%

Dodavatel: Apollo Scientific

OR960202-1G OR960202-5G
APOSOR960202-1GEA 2360 CZK
APOSOR960202-1G APOSOR960202-5G
6-nitro-3,4-dihydro-2H-1,4-benzoxazine 98%
6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Vzorec: C₈H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD07698591
CAS číslo: 28226-22-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR