Tisk…

2-Bromo-5-iodoanisole 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-4-iodo-2-methoxybenzene

OR9601-5G OR9601-1G OR9601-25G
APOSOR9601-5GEA 1240 CZK
APOSOR9601-5G APOSOR9601-1G APOSOR9601-25G
2-Bromo-5-iodoanisole 98%
2-Bromo-5-iodoanisole
Vzorec: C₇H₆BrIO
MW: 312,93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07780651
CAS číslo: 755027-18-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR