Tisk…

2H-1,4-Benzoxazine-2,3(4H)-dione 98%

Dodavatel: Apollo Scientific

Synonyms: 3-Hydroxy-2H-benzo[b][1,4]oxazin-2-one

OR959825-1G OR959825-5G
APOSOR959825-1GEA 6090 CZK
APOSOR959825-1G APOSOR959825-5G
2H-1,4-Benzoxazine-2,3(4H)-dione 98%
2H-1,4-Benzoxazine-2,3(4H)-dione
Vzorec: C₈H₅NO₃
MW: 163,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00584446
CAS číslo: 3597-63-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR