2,6-Dimethylbenzothiazole 95%
Dodavatel: Apollo Scientific
OR959556-1G
OR959556-5G
APOSOR959556-1GEA
4570
CZK
APOSOR959556-1G
APOSOR959556-5G
2,6-Dimethylbenzothiazole 95%
2,6-Dimethylbenzothiazole
Vzorec:
C₉H₉NS Storage Temperature: Ambient |
CAS číslo:
2941-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...