Tisk…

5-Methoxy-2-methylbenzothiazole 95%

Dodavatel: Apollo Scientific

OR959552-1G OR959552-5G
APOSOR959552-1GEA 3540 CZK
APOSOR959552-1G APOSOR959552-5G
5-Methoxy-2-methylbenzothiazole 95%
5-Methoxy-2-methylbenzothiazole
Vzorec: C₉H₉NOS
MW: 179,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082664
CAS číslo: 2941-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR