Tisk…

Cyclopentyl-2-pyridyl ketone 98%

Dodavatel: Apollo Scientific

OR959518-1G OR959518-5G
APOSOR959518-1GEA 3710 CZK
APOSOR959518-1G APOSOR959518-5G
Cyclopentyl-2-pyridyl ketone 98%
Cyclopentyl-2-pyridyl ketone
Vzorec: C₁₁H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD07699202
CAS číslo: 157592-43-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR