4-(2-Chloropyrimidin-4-yl)morpholine 96%
Dodavatel: Apollo Scientific
OR959389-1G
OR959389-5G
APOSOR959389-1GEA
4470
CZK
APOSOR959389-1G
APOSOR959389-5G
4-(2-Chloropyrimidin-4-yl)morpholine 96%
4-(2-Chloropyrimidin-4-yl)morpholine
Vzorec:
C₈H₁₀ClN₃O Storage Temperature: Ambient |
MDL Number:
MFCD00981910 CAS číslo: 62968-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...