Tisk…

Methyl-5-aminopicolinate 96%

Dodavatel: Apollo Scientific

Synonyms: Methyl 5-aminopyridine-2-carboxylate, Methyl 5-aminopicolinate

OR959381-10G OR959381-1G OR959381-5G
APOSOR959381-10GEA 7050 CZK
APOSOR959381-10G APOSOR959381-1G APOSOR959381-5G
Methyl-5-aminopicolinate 96%
Methyl-5-aminopicolinate
Vzorec: C₇H₈N₂O₂
MW: 152,1506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783939
CAS číslo: 67515-76-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR