Tisk…

3-(2-Methoxyethoxy)aniline 95%

Dodavatel: Apollo Scientific

OR959268-1G OR959268-250MG
APOSOR959268-1GEA 13860 CZK
APOSOR959268-1G APOSOR959268-250MG
3-(2-Methoxyethoxy)aniline 95%
3-(2-Methoxyethoxy)aniline
Vzorec: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD08691445
CAS číslo: 110178-35-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR