Tisk…

2,6-Dimethyl-4-heptanol 95%

Dodavatel: Apollo Scientific

Synonyms: Diisobutylcarbinol

OR959119-25G OR959119-100G OR959119-5G
APOSOR959119-25GEA 3640 CZK
APOSOR959119-25G APOSOR959119-100G APOSOR959119-5G
2,6-Dimethyl-4-heptanol 95%
2,6-Dimethyl-4-heptanol
Vzorec: C₉H₂₀O
MW: 144.26
Bod varu: 173
Bod tání: -65
Storage Temperature: Ambient
MDL Number: MFCD00008944
CAS číslo: 108-82-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR